5-Amino-3-methyl-1,2,4-thiadiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009666-1G 009666-5G
FLUO009666-1GEA 0 GBP
FLUO009666-1G FLUO009666-5G
5-Amino-3-methyl-1,2,4-thiadiazole
5-Amino-3-methyl-1,2,4-thiadiazole
Formula: C₃H₅N₃S
MW: 115.16 g/mol
MDL Number: MFCD01114751
CAS Number: 17467-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR