3,5-Dichloro-1,2,4-thiadiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005950-5G 05950.1G 005950-1G
FLUO005950-5GEA 0 GBP
FLUO005950-5G FLUO05950.1G FLUO005950-1G
3,5-Dichloro-1,2,4-thiadiazole
3,5-Dichloro-1,2,4-thiadiazole
Formula: C₂Cl₂N₂S
MW: 155.01 g/mol
MDL Number: MFCD00799286
CAS Number: 2254-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR