2-Bromo-1,3,4-thiadiazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1492-5G
HI-1492-1G
COBBHI-1492-5GEA
0
GBP
COBBHI-1492-5G
COBBHI-1492-1G
2-Bromo-1,3,4-thiadiazole
2-Bromo-1,3,4-thiadiazole
Formula:
C₂HBrN₂S |
MDL Number:
MFCD04115375 CAS Number: 61929-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...