3-Methylbenzo[b]thiophene-2-carboxaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
023335-5G
023335-1G
FLUO023335-5GEA
0
GBP
FLUO023335-5G
FLUO023335-1G
3-Methylbenzo[b]thiophene-2-carboxaldehyde
3-Methylbenzo[b]thiophene-2-carboxaldehyde
Formula:
C₁₀H₈OS MW: 176.24 g/mol |
MDL Number:
MFCD00052307 CAS Number: 22053-74-3 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...