3-Methylbenzo[b]thiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Methylbenzothiophene

018819-1G 018819-5G
FLUO018819-1GEA 0 GBP
FLUO018819-1G FLUO018819-5G
3-Methylbenzo[b]thiophene
3-Methylbenzo[b]thiophene
Formula: C₉H₈S
MW: 148.23 g/mol
MDL Number: MFCD00040243
CAS Number: 1455-18-1
EINECS: 215-934-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR