1,3-Diethynylbenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009134-5G 009134-25G
FLUO009134-5GEA 0 GBP
FLUO009134-5G FLUO009134-25G
1,3-Diethynylbenzene
1,3-Diethynylbenzene
Formula: C₁₀H₆
MDL Number: MFCD00041686
CAS Number: 1785-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR