2,2,6,6-Tetramethyl-4-piperidone monohydrate ≥99%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Triacetonamine monohydrate, 2,2,6,6-Tetramethyl-4-piperidone Hydrate

A16211.09 A16211.18 A16211.30
A16211.09EA 26 GBP
A16211.09 A16211.18 A16211.30
2,2,6,6-Tetramethyl-4-piperidone monohydrate ≥99%
2,2,6,6-Tetramethyl-4-piperidone monohydrate

Purity: 99% CAS No.: 10581-38-1 Molecular Formula: C9H17NO.H2O Formula Weight: 173.26 (155.24anhy) Storage Temperature: 2°C to 8°C Melting Point: 63-61° MDL No.: MFCD00149393

Formula: C₉H₁₇NO·1H₂O
MW: 173.26 g/mol
Melting Pt: 63…61 °C
MDL Number: MFCD00149393
CAS Number: 10581-38-1
EINECS: 212-554-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR