5-Bromobenzo[b]thiophene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromobenzothiophene

HI-1677-5G HI-1677-1G
COBBHI-1677-5GEA 0 GBP
COBBHI-1677-5G COBBHI-1677-1G
5-Bromobenzo[b]thiophene
5-Bromobenzo[b]thiophene
Formula: C₈H₅BrS
MW: 213.1 g/mol
MDL Number: MFCD03069318
CAS Number: 4923-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR