Thiazolidine-2-carboxylic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005324.5G
005324-25G
005324-100G
FLUO005324.5GEA
58
GBP
FLUO005324.5G
FLUO005324-25G
FLUO005324-100G
Thiazolidine-2-carboxylic acid
Thiazolidine-2-carboxylic acid
Formula:
C₄H₇NO₂S |
MDL Number:
MFCD00068005 CAS Number: 65126-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...