2,4-Thiazolidinedione

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Thiazolidine-2,4-dione

HI-1209-100G HI-1209-25G
COBBHI-1209-100GEA 0 GBP
COBBHI-1209-100G COBBHI-1209-25G
2,4-Thiazolidinedione
2,4-Thiazolidinedione
Formula: C₃H₃NO₂S
MW: 117.13 g/mol
MDL Number: MFCD00005478
CAS Number: 2295-31-0
EINECS: 218-941-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR