(Phenylthio)acetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003773-5G 003773-25G
FLUO003773-5GEA 0 GBP
FLUO003773-5G FLUO003773-25G
(Phenylthio)acetonitrile
(Phenylthio)acetonitrile
Formula: C₈H₇NS
MDL Number: MFCD00019831
CAS Number: 5219-61-4
EINECS: 226-013-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR