α-Ketoglutaric acid monosodium salt, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ300

SIALK1875-1KGEA 3030 GBP
SIALK1875-1KG SIALK1875-25G SIALK1875-100G SIALK1875-1G SIALK1875-5G
α-Ketoglutaric acid monosodium salt, Sigma-Aldrich®
α-Ketoglutaric acid monosodium salt
Formula: C₅H₅NaO₅
MW: 168.08 g/mol
MDL Number: MFCD00064196
CAS Number: 22202-68-2
EINECS: 244-836-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR