α-Ketoglutaric acid monopotassium salt, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Potassium 4-carboxy-2-oxobutanoate
M-Clarity™ quality level = MQ300
K2000-5G
K2000-100G
K2000-1KG
K2000-25G
SIALK2000-5GEA
54.3
GBP
SIALK2000-5G
SIALK2000-100G
SIALK2000-1KG
SIALK2000-25G
α-Ketoglutaric acid monopotassium salt, Sigma-Aldrich®
α-Ketoglutaric acid monopotassium salt
Formula:
C₅H₅KO₅ MW: 184.19 g/mol |
MDL Number:
MFCD00064195 CAS Number: 997-43-3 EINECS: 213-641-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...