Allyl chloroformate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
242306-5G
242306-100G
SIAL242306-5GEA
51.5
GBP
SIAL242306-5G
SIAL242306-100G
Allyl chloroformate, Sigma-Aldrich®
Allyl chloroformate
Formula:
C₄H₅ClO₂ |
MDL Number:
MFCD00000648 CAS Number: 2937-50-0 EINECS: 220-916-6 UN: 1722 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...