4-Chloro-2,6-diaminopyrimidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4-Diamino-6-chloropyrimidine, 6-Chloropyrimidine-2,4-diamine

011624-100G 011624-25G
FLUO011624-100GEA 0 GBP
FLUO011624-100G FLUO011624-25G
4-Chloro-2,6-diaminopyrimidine
4-Chloro-2,6-diaminopyrimidine
Formula: C₄H₅ClN₄
MW: 144.56 g/mol
MDL Number: MFCD00006097
CAS Number: 156-83-2
EINECS: 205-863-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR