3-Amino-1-methyl-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methyl-1H-pyrazol-3-amine, 3-Amino-1-methylpyrazole, 1-Methyl-1H-pyrazol-3-ylamine, 3-Amino-1-methyl-1H-pyrazole

HI-1414-5G HI-1414-25G HI-1414-1G
COBBHI-1414-5GEA 0 GBP
COBBHI-1414-5G COBBHI-1414-25G COBBHI-1414-1G
3-Amino-1-methyl-1H-pyrazole
3-Amino-1-methyl-1H-pyrazole
Formula: C₄H₇N₃
MW: 97.12 g/mol
MDL Number: MFCD00466340
CAS Number: 1904-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR