3-Aminocoumarin 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Amino-2H-chromen-2-one

L14031.MC L14031.ME L14031.04
L14031.MCEA 20.7 GBP
L14031.MC L14031.ME L14031.04
3-Aminocoumarin 97%
3-Aminocoumarin

Purity: 97% CAS No.: 1635-31-0 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: 135-137° Beilstein Reference: 128033

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00016965
CAS Number: 1635-31-0
EINECS: 216-659-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR