Denatonium benzoate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-Benzyl-2-((2,6-dimethylphenyl)amino)-N,N-diethyl-2-oxoethanaminium benzoate
M-Clarity™ quality level = MQ100
D5765-10G
D5765-1G
D5765-5G
SIALD5765-10GEA
292
GBP
SIALD5765-10G
SIALD5765-1G
SIALD5765-5G
Denatonium benzoate, Sigma-Aldrich®
Denatonium benzoate
Formula:
C₂₈H₃₄N₂O₃ |
MDL Number:
MFCD00031578 CAS Number: 3734-33-6 EINECS: 223-095-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...