2,5-Dichlorohydroquinone ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

B21285.03 B21285.06
B21285.03EA 30.9 GBP
B21285.03 B21285.06
2,5-Dichlorohydroquinone ≥97%
2,5-Dichlorohydroquinone

Purity: 97% CAS No.: 824-69-1 Molecular Formula: C6H4Cl2O2 Formula Weight: 179.00 Melting Point: 168-171° MDL No.: MFCD00041749

Formula: C₆H₄Cl₂O₂
MW: 179 g/mol
Melting Pt: 168…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00041749
CAS Number: 824-69-1
EINECS: 212-533-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR