Ethyl oxalyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl oxalyl monochloride, Oxalic Acid Monoethyl Ester Chloride, Ethyl Chlorooxoacetate, Ethyl Chloroglyoxylate, Chloroglyoxylic Acid Ethyl Ester, Ethyl chloroformylformate

024489-1KG 024489-100G
FLUO024489-1KGEA 0 GBP
FLUO024489-1KG FLUO024489-100G
Ethyl oxalyl chloride
Ethyl oxalyl chloride
Formula: C₄H₅ClO₃
MDL Number: MFCD00000706
CAS Number: 4755-77-5
EINECS: 225-285-0
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR