Vinyltrimethylsilane
Supplier: ABCR GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Trimethyl(vinyl)silane
ABCRAB109192.0010EA
0
GBP
ABCRAB109192.0010
ABCRAB109192.0025
Vinyltrimethylsilane
Vinyltrimethylsilane
Formula:
C₅H₁₂Si MW: 100.24 g/mol Boiling Pt: 55 °C (1013 hPa) Density: 0.7038 g/cm³ (20 °C) |
MDL Number:
MFCD00008606 CAS Number: 754-05-2 EINECS: 212-042-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...