(±)-Ethyl-2-chloropropionate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

192414-1L 192414-25ML
SIAL192414-1LEA 275 GBP
SIAL192414-1L SIAL192414-25ML
(±)-Ethyl-2-chloropropionate, Sigma-Aldrich®
(±)-Ethyl 2-chloropropionate
Formula: C₅H₉ClO₂
MW: 136.58 g/mol
MDL Number: MFCD00000864
CAS Number: 535-13-7
EINECS: 208-610-0
UN: 2935
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR