4-Bromo-6-tert-butyl-2-mercaptobenzothiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1622-1G HI-1622-5G
COBBHI-1622-1GEA 0 GBP
COBBHI-1622-1G COBBHI-1622-5G
4-Bromo-6-tert-butyl-2-mercaptobenzothiazole
4-Bromo-6-tert-butyl-2-mercaptobenzothiazole
Formula: C₁₁H₁₂BrNS₂
MW: 302.26 g/mol
MDL Number: MFCD13195713
CAS Number: 1199773-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR