1-Bromo-4-phenylbutane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008935-1G 008935-5G
FLUO008935-1GEA 0 GBP
FLUO008935-1G FLUO008935-5G
1-Bromo-4-phenylbutane
1-Bromo-4-phenylbutane
Formula: C₁₀H₁₃Br
MDL Number: MFCD00154988
CAS Number: 13633-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR