2,4-Dimethylthiophene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013555-5G
013555-1G
FLUO013555-5GEA
0
GBP
FLUO013555-5G
FLUO013555-1G
2,4-Dimethylthiophene
2,4-Dimethylthiophene
Formula:
C₆H₈S MW: 112.2 g/mol |
MDL Number:
MFCD00130082 CAS Number: 638-00-6 EINECS: 211-312-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...