3-Methylthiophene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003448-100G
003448-25G
FLUO003448-100GEA
0
GBP
FLUO003448-100G
FLUO003448-25G
3-Methylthiophene
3-Methylthiophene
Formula:
C₅H₆S MW: 98.17 g/mol Boiling Pt: ∼114 °C (1013 hPa) Melting Pt: –69 °C Density: 1.02 g/cm³ (20 °C) Flash Pt: 11 Storage Temperature: Ambient |
MDL Number:
MFCD00005470 CAS Number: 616-44-4 EINECS: 210-482-6 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...