2-Methylthiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

001209-100G 001209-25G
FLUO001209-100GEA 0 GBP
FLUO001209-100G FLUO001209-25G
2-Methylthiophene
Formula: C₅H₆S
Boiling Pt: 113 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005451
CAS Number: 554-14-3
EINECS: 209-063-0
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR