2,5-Diiodothiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001076-5G 001076-25G
FLUO001076-5GEA 0 GBP
FLUO001076-5G FLUO001076-25G
2,5-Diiodothiophene
2,5-Diiodothiophene
Formula: C₄H₂I₂S
Boiling Pt: 67…70 °C (0.05 mmHg)
Melting Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014525
CAS Number: 625-88-7
EINECS: 210-915-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR