3-Bromothiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Thienyl bromide

003082-100G 003082-25G 003082-500G
FLUO003082-100GEA 0 GBP
FLUO003082-100G FLUO003082-25G FLUO003082-500G
3-Bromothiophene
3-Bromothiophene
Formula: C₄H₃BrS
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005464
CAS Number: 872-31-1
EINECS: 212-821-3
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR