3,4-Dibromothiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001057-5G 001057-25G
FLUO001057-5GEA 0 GBP
FLUO001057-5G FLUO001057-25G
3,4-Dibromothiophene
3,4-Dibromothiophene
Formula: C₄H₂Br₂S
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: 4,5 °C
Density: 2,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005465
CAS Number: 3141-26-2
EINECS: 221-546-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR