2-Bromothiophene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001055-50G
001055-1KG
001055-250G
FLUO001055-50GEA
0
GBP
FLUO001055-50G
FLUO001055-1KG
FLUO001055-250G
2-Bromothiophene
2-Bromothiophene
Formula:
C₄H₃BrS Boiling Pt: ∼149 °C (1013 hPa) Melting Pt: –10 °C Density: 1.71 g/cm³ (20 °C) Flash Pt: 52 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005417 CAS Number: 1003-09-4 EINECS: 213-699-4 UN: 2929 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...