2,5-Dibromothiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001056-250G 001056-50G
FLUO001056-250GEA 0 GBP
FLUO001056-250G FLUO001056-50G
2,5-Dibromothiophene
2,5-Dibromothiophene
Formula: C₄H₂Br₂S
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: –6 °C
Density: 2.1531 g/cm³ (13.8 °C)
MDL Number: MFCD00005420
CAS Number: 3141-27-3
EINECS: 221-547-3
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR