2,3,5-Tribromothiophene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001452-1G
001452-10G
001452-50G
FLUO001452-1GEA
0
GBP
FLUO001452-1G
FLUO001452-10G
FLUO001452-50G
2,3,5-Tribromothiophene
2,3,5-Tribromothiophene
Formula:
C₄HBr₃S MW: 320.83 g/mol Boiling Pt: 80 °C (0.5 mmHg) Melting Pt: 29 °C |
MDL Number:
MFCD00014521 CAS Number: 3141-24-0 EINECS: 221-544-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...