2,3,4-Tribromothiophene ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

L03946.03 L03946.06
L03946.03EA 43 GBP
L03946.03 L03946.06
2,3,4-Tribromothiophene ≥97%
2,3,4-Tribromothiophene

Purity: 97% CAS No.: 3141-25-1 Molecular Formula: C4HBr3S Formula Weight: 320.84 Melting Point: 43-45° Boiling Point: 150-152°/20mm UN No.: 2811 MDL No.: MFCD00041210

Formula: C₄HBr₃S
Boiling Pt: 150…152 °C (20 mmHg)
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00041210
CAS Number: 3141-25-1
EINECS: 221-545-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR