3-(Trifluoromethyl)pyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007663-1G
007663-5G
007663-10G
FLUO007663-1GEA
0
GBP
FLUO007663-1G
FLUO007663-5G
FLUO007663-10G
3-(Trifluoromethyl)pyridine
3-(Trifluoromethyl)pyridine
Formula:
C₆H₄F₃N Boiling Pt: 110…113 °C (1013 hPa) |
MDL Number:
MFCD00100032 CAS Number: 3796-23-4 EINECS: 223-268-2 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...