3-Iodopyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

008065-5G 008065-25G
FLUO008065-5GEA 0 GBP
FLUO008065-5G FLUO008065-25G
3-Iodopyridine
3-Iodopyridine
Formula: C₅H₄IN
Boiling Pt: 90…92 °C (14 mmHg)
Melting Pt: 53.5 °C
MDL Number: MFCD00023553
CAS Number: 1120-90-7
EINECS: 214-322-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR