2-Fluoropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

001297-25G 001297-100G 001297-250G
FLUO001297-25GEA 0 GBP
FLUO001297-25G FLUO001297-100G FLUO001297-250G
2-Fluoropyridine
2-Fluoropyridine
Formula: C₅H₄FN
Boiling Pt: 126 °C (1013 hPa)
Density: 1.129 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006224
CAS Number: 372-48-5
EINECS: 206-757-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR