2,6-Difluoropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

001607-100G 001607-25G
FLUO001607-100GEA 0 GBP
FLUO001607-100G FLUO001607-25G
2,6-Difluoropyridine
2,6-Difluoropyridine
Formula: C₅H₃F₂N
Boiling Pt: 123…124 °C (1013 hPa)
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006227
CAS Number: 1513-65-1
EINECS: 216-152-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR