3,5-Dichloro-2,4,6-trifluoropyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3,5-Dichlorotrifluoropyridine
004752-5G
004752-25G
004752-100G
FLUO004752-5GEA
0
GBP
FLUO004752-5G
FLUO004752-25G
FLUO004752-100G
3,5-Dichloro-2,4,6-trifluoropyridine
3,5-Dichloro-2,4,6-trifluoropyridine
Formula:
C₅Cl₂F₃N MW: 201.96 g/mol Boiling Pt: 159…160 °C (1013 hPa) Melting Pt: 24…26 °C Density: 1.62 g/cm³ (20 °C) |
MDL Number:
MFCD00006226 CAS Number: 1737-93-5 EINECS: 217-088-3 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...