2,5-Dibromopyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
019467-100G
019467-25G
FLUO019467-100GEA
0
GBP
FLUO019467-100G
FLUO019467-25G
2,5-Dibromopyridine
2,5-Dibromopyridine
Formula:
C₅H₃Br₂N Boiling Pt: 234…235 °C (1013 hPa) Melting Pt: 90…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006221 CAS Number: 624-28-2 EINECS: 210-839-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...