Silver fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002862-5G 002862-10G 002862-50G
FLUO002862-5GEA 0 GBP
FLUO002862-5G FLUO002862-10G FLUO002862-50G
Silver fluoride
Silver fluoride
Formula: AgF
Boiling Pt: 1150 °C (1013 hPa)
Melting Pt: 433 °C
Density: 5.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003410
CAS Number: 7775-41-9
EINECS: 231-895-8
UN: 3260
ADR: 8,II
Merck Index: 12,08657

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR