Lanthanum(III) fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Lanthanum trifluoride

006706-25G 006706-100G
FLUO006706-25GEA 0 GBP
FLUO006706-25G FLUO006706-100G
Lanthanum(III) fluoride
Lanthanum (III) fluoride
Formula: F₃La
Boiling Pt: ∼2327 °C (1013 hPa)
Melting Pt: 1491.85…1507.85 °C
Density: 5.936 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011073
CAS Number: 13709-38-1
EINECS: 237-252-8
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR