1-Isoquinolinecarboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Isoquinoline-1-carboxylic acid

019060-1G 019060-5G 019060-25G
FLUO019060-1GEA 0 GBP
FLUO019060-1G FLUO019060-5G FLUO019060-25G
1-Isoquinolinecarboxylic acid
1-Isoquinolinecarboxylic acid
Formula: C₁₀H₇NO₂
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD00006901
CAS Number: 486-73-7
EINECS: 207-639-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR