1,3-Indandione
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
011171-25G
011171-100G
FLUO011171-25GEA
0
GBP
FLUO011171-25G
FLUO011171-100G
1,3-Indandione
1,3-Indandione
Formula:
C₉H₆O₂ Boiling Pt: 283 °C (1013 hPa) Melting Pt: 129…132 °C Density: 1.37 g/cm³ (21 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003779 CAS Number: 606-23-5 EINECS: 210-109-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...