Phosphorus(V) chloride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
79590-500G
79590-50G
79590-1KG
SIAL79590-500GEA
73.5
GBP
SIAL79590-500G
SIAL79590-50G
SIAL79590-1KG
Phosphorus(V) chloride, Sigma-Aldrich®
Phosphorus (V) chloride
Formula:
Cl₅P MW: 208.24 g/mol Melting Pt: ∼148 °C Density: 3.6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011439 CAS Number: 10026-13-8 EINECS: 233-060-3 UN: 1806 ADR: 8,II Merck Index: 12,07508 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...