Carbon disulphide ≥99.9%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
32472.AKEA
38.2
GBP
32472.AK
32472.K2
Carbon disulphide ≥99.9%
Carbon disulphide
Formula:
CS₂ MW: 76.14 g/mol Boiling Pt: 46 °C Melting Pt: –112 °C Density: 1.2632 Flash Pt: –30 °C (–22 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011321 CAS Number: 75-15-0 EINECS: 200-843-6 UN: 1131 ADR: 3,I Merck Index: 14,1811 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...