Phenylphosphonic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Phenylphosphonic Acid
M-Clarity™ quality level = MQ200
P29006-100G
P29006-500G
P29006-5G
SIALP29006-100GEA
70
GBP
SIALP29006-100G
SIALP29006-500G
SIALP29006-5G
Phenylphosphonic acid, Sigma-Aldrich®
Phenylphosphonic acid
Formula:
C₆H₇O₃P MW: 158.09 g/mol |
MDL Number:
MFCD00002136 CAS Number: 1571-33-1 EINECS: 216-388-1 UN: 3261 ADR: 8,III |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...