N,N-Dimethylglycine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

D1156-10G D1156-10MG D1156-25G D1156-5G
SIALD1156-10GEA 209 GBP
SIALD1156-10G SIALD1156-10MG SIALD1156-25G SIALD1156-5G
N,N-Dimethylglycine, Sigma-Aldrich®
N,N-Dimethylglycine
Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00004283
CAS Number: 1118-68-9
EINECS: 214-267-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR