Potassium fluoride on aluminium oxide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
316385-100G
316385-25G
SIAL316385-100GEA
368
GBP
SIAL316385-100G
SIAL316385-25G
Potassium fluoride on aluminium oxide, Sigma-Aldrich®
Potassium fluoride
Formula:
KF MW: 58.1 g/mol Boiling Pt: 1505 Melting Pt: 860 |
MDL Number:
MFCD00011398 CAS Number: 7789-23-3 UN: 1812 ADR: 6.1 III Merck Index: 13,07714 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...