Ethylphosphonic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
289876-1G
289876-5G
SIAL289876-1GEA
73.9
GBP
SIAL289876-1G
SIAL289876-5G
Ethylphosphonic acid, Sigma-Aldrich®
Ethylphosphonic acid
Formula:
C₂H₇O₃P |
MDL Number:
MFCD00002141 CAS Number: 6779-09-5 EINECS: 229-839-2 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...